4-Aminobutyric acid, 99+%
4-Aminobutyric acid, 99+%
4-Aminobutyric acid, 99+%
Thermo Scientific Chemicals

4-Aminobutyric acid, 99+%

CAS: 56-12-2 | C4H9NO2 | 103.121 g/mol
Have Questions?
Catalog NumberQuantity
J61307.09
also known as J61307-09
10 g
Catalog number J61307.09
also known as J61307-09
Price (EUR)
26,70
Each
Quantity:
10 g
Request bulk or custom format
Price (EUR)
26,70
Each
Chemical Identifiers
CAS56-12-2
IUPAC Name4-aminobutanoic acid
Molecular FormulaC4H9NO2
InChI KeyBTCSSZJGUNDROE-UHFFFAOYSA-N
SMILESNCCCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Residue on ignition0.7% max.
Appearance (Color)White
pH6.5 - 7.5
Assay (unspecified)99.0% min.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only