TES, 0.2M buffer soln., pH 7.5
TES, 0.2M buffer soln., pH 7.5
TES, 0.2M buffer soln., pH 7.5
Thermo Scientific Chemicals

TES, 0.2M buffer soln., pH 7.5

CAS: 70331-82-7 | C6H14NNaO6S | 251.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
250 mL
Catalog number J61350.AK
also known as J61350-AK
Price (EUR)
130,00
Each
Quantity:
250 mL
Request bulk or custom format
Price (EUR)
130,00
Each
Chemical Identifiers
CAS70331-82-7
IUPAC Namesodium 2-{[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino}ethane-1-sulfonate
Molecular FormulaC6H14NNaO6S
InChI KeyGUXMFAHOYNRHBI-UHFFFAOYSA-M
SMILES[Na+].OCC(CO)(CO)NCCS([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
CompositionTES: 200 mM
pH7.35-7.65
FormLiquid
TES, 0.2M buffer soln., is a suitable buffer for epidermal cell growth at pH 7.4 - 7.9, and is also useful for studying succinate oxidation. It is also widely used as a buffer in biological applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
TES, 0.2M buffer soln., is a suitable buffer for epidermal cell growth at pH 7.4 - 7.9, and is also useful for studying succinate oxidation. It is also widely used as a buffer in biological applications.

Solubility
Soluble in water (350 g/L) at 20°C, and methanol(very faint turbidity).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. NE Good.; GD Winget.; W Winter.; TN Connolly. Hydrogen ion buffers for biological research. Biochemistry. 1966, 5, (2), 467-477.
  2. S Matsushita. Coloring conditions of thiobarbituric acid test for detecting lipid hydro peroxides. Lipids. 1980, 15, (3), 137-140.