Soybean oil
Thermo Scientific Chemicals

Soybean oil

CAS: 8001-22-7
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
Catalog number J61399.AP
also known as J61399-AP
Price (EUR)
40,10
Each
Quantity:
500 mL
Request bulk or custom format
Price (EUR)
40,10
Each
Chemical Identifiers
CAS8001-22-7
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to pale yellow
FormOily liquid
Peroxide content≤10.0 meq/kg
Refractive Index1.4650-1.4750 @ 20°C
Saponification ValueUnsaponifiable matter: ≤1.5%
Soybean oil is used as a base for printing inks and oil paints. It is also used as a fixative to extend the short duration of action of essential oils such as geranium oil in several commercial products. Further, it is used on citrus fruits and domestic dwellings. It is also employed in the determination of freezing point of soybean oil. It plays as a delivery vehicle of alfa-lipoic acid. In addition to this, it is used as a condiment for salads as well as involved in the synthesis of epoxidized-soybean oil (ESO).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Soybean oil is used as a base for printing inks and oil paints. It is also used as a fixative to extend the short duration of action of essential oils such as geranium oil in several commercial products. Further, it is used on citrus fruits and domestic dwellings. It is also employed in the determination of freezing point of soybean oil. It plays as a delivery vehicle of alfa-lipoic acid. In addition to this, it is used as a condiment for salads as well as involved in the synthesis of epoxidized-soybean oil (ESO).

Solubility
Miscible with absolute alcohol, ether, petroleum ether. Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Ferrero, G. O.; Almeida, M. F.; Alvim-Ferraz, M. C. M.; Dias, J. M. Glycerol-enriched heterogeneous catalyst for biodiesel production from soybean oil and waste frying oil. Energy Convers. Manage. 2015, 89, 665-671.
  2. Luna, A. S.; da Silva, A. P.; Pinho, J. S. A.; Ferré, J.; Boqué, R. A novel approach to discriminate transgenic from non-transgenic soybean oil using FT-MIR and chemometrics. Food Res. Int. 2015, 67, 206-211.