N-Succinimidyl 3-maleimidobenzoate, 95%
N-Succinimidyl 3-maleimidobenzoate, 95%
N-Succinimidyl 3-maleimidobenzoate, 95%
Thermo Scientific Chemicals

N-Succinimidyl 3-maleimidobenzoate, 95%

A heterobifunctional coupling reagent useful for forming enzyme immunoconjugates | CAS: 58626-38-3 | C15H10N2O6 | 314.253 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number J61410.03
also known as J61410-03
Price (EUR)
260,00
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
260,00
Each
Specifications
Beilstein1505254
CAS58626-38-3
Chemical Name or MaterialN-Succinimidyl 3-maleimidobenzoate
ColorWhite
EINECS Number261-368-8
View more
N-Succinimidyl 3-maleimidobenzoate is useful for forming enzyme immunoconjugates. It has a 9.9 Angstrom spacer arm. Heterobifunctional crosslinking reagent reactive toward primary amine and sulfhydryl. Useful for preparation of enzyme immunoconjugates and hapten carrier molecule conjugates

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Succinimidyl 3-maleimidobenzoate is useful for forming enzyme immunoconjugates. It has a 9.9 Angstrom spacer arm. Heterobifunctional crosslinking reagent reactive toward primary amine and sulfhydryl. Useful for preparation of enzyme immunoconjugates and hapten carrier molecule conjugates

Solubility
Soluble in DMF or ethyl acetate.

Notes
Moisture & Light Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

General References:

  1. J. Fiserova.; K. Schwarzerova.; J. Petrasek.; Z. Opatrny. ARP2 and ARP3 are localized to sites of actin filament nucleation in tobacco BY-2 cells. Protoplasma. 2006, 227, (2), 119-128.
  2. Lisa C. Shriver-Lake.; Brian Donner.; Rebecca Edelstein.; Kristen Breslin.; Suresh K. Bhatia.; Frances S. Ligler. Antibody immobilization using heterobifunctional crosslinkers. Biosensors and Bioelectronics. 1997, 12, (11), 1101-1106.