(+/-)-Verapamil hydrochloride, 99+%
(+/-)-Verapamil hydrochloride, 99+%
(+/-)-Verapamil hydrochloride, 99+%
Thermo Scientific Chemicals

(+/-)-Verapamil hydrochloride, 99+%

CAS: 152-11-4 | C27H39ClN2O4 | 491.069 g/mol
Have Questions?
Catalog NumberQuantity
J61535.03
also known as J61535-03
1 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J61535.03
also known as J61535-03
Price (EUR)
68,65
Online exclusive
75,60
Save 6,95 (9%)
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
68,65
Online exclusive
75,60
Save 6,95 (9%)
Each
Chemical Identifiers
CAS152-11-4
IUPAC Namehydrogen 2-(3,4-dimethoxyphenyl)-5-{[2-(3,4-dimethoxyphenyl)ethyl](methyl)amino}-2-(propan-2-yl)pentanenitrile chloride
Molecular FormulaC27H39ClN2O4
InChI KeyDOQPXTMNIUCOSY-UHFFFAOYNA-N
SMILES[H+].[Cl-].COC1=CC=C(CCN(C)CCCC(C#N)(C(C)C)C2=CC=C(OC)C(OC)=C2)C=C1OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormPowder or crystalline powder
Assay (Titration ex Chloride)≥99.0 to ≤101.0% (non-U.S. specification)
Assay≥99.0% (U.S. specification)
CommentSpecification differs for U.S. and non-U.S. material where indicated
±)-Verapamil hydrochloride is a calcium channel modulator, adrenoceptor antagonist, anti-arrhythmic, cardiac depressant, and coronary vasodilator. It is also a calcium channel-blocker. It acts by inhibiting the slow channel entry of calcium into the cell. It acts by plugging up the channels and limiting the entry of calcium into both smooth muscle cells of arteriolar walls and the cardiac muscle cells at higher doses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
±)-Verapamil hydrochloride is a calcium channel modulator, adrenoceptor antagonist, anti-arrhythmic, cardiac depressant, and coronary vasodilator. It is also a calcium channel-blocker. It acts by inhibiting the slow channel e
RUO – Research Use Only