Clozapine, 97%
Clozapine, 97%
Clozapine, 97%
Thermo Scientific Chemicals

Clozapine, 97%

Potent, selective muscarinic antagonist | CAS: 5786-21-0 | C18H19ClN4 | 326.828 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mg
100 mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J61583.MB
also known as J61583-MB
Price (EUR)
95,65
Online Exclusive
106,00
Save 10,35 (10%)
Each
Quantity:
25 mg
Request bulk or custom format
Price (EUR)
95,65
Online Exclusive
106,00
Save 10,35 (10%)
Each
Specifications
CAS5786-21-0
Chemical Name or MaterialClozapine
ColorYellow
DOT InformationTransport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECS Number227-313-7
View more
Potent, selective muscarinic antagonist

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only