m-Tolyl acetate, 97%
m-Tolyl acetate, 97%
m-Tolyl acetate, 97%
Thermo Scientific Chemicals

m-Tolyl acetate, 97%

CAS: 111072-31-2 | C22H16N7NaO13S2 | 673.52 g/mol
Have Questions?
Catalog NumberQuantity
J61726.MD250 mg
Catalog number J61726.MD
Price (EUR)
381,00
Each
Quantity:
250 mg
Request bulk or custom format
Price (EUR)
381,00
Each
Chemical Identifiers
CAS111072-31-2
IUPAC Namesodium 2,3-bis(2-methoxy-4-nitro-5-sulfonatophenyl)-5-(phenylcarbamoyl)-3H-1,2λ⁵,3,4-tetrazol-2-ylium
Molecular FormulaC22H16N7NaO13S2
InChI KeyJACYMBNQPPWQML-UHFFFAOYSA-M
SMILES[Na+].COC1=C(C=C(C(=C1)[N+]([O-])=O)S([O-])(=O)=O)N1N=C(N=[N+]1C1=C(OC)C=C(C(=C1)S([O-])(=O)=O)[N+]([O-])=O)C(=O)NC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)Yellow
Proton NMRConforms to structure
Loss on Drying≤15.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Tetrazolium derivative useful in cancer research.XTT sodium salt is used in conjunction with phenazine methosulfate to screen human cancer cell lines. It is also used to study fungal cell damage. It is involved in testing antimicrob
RUO – Research Use Only

General References:

  1. Chew, S. Y.; Cheah, Y. K.; Seow, H. F.; Sandai, D.; Than, L. T. L. In vitro Modulation of Probiotic Bacteria on the Biofilm of Candida glabrata. Anaerobe 2015, 34, 132-138.
  2. Khurram, M.; Lawton, L. A.; Edwards, C.; Iriti, M.; Hameed, A.; Khan, M. A.; Khan, F. A.; Rahman, S. U. Rapid Bioassay-Guided Isolation of Antibacterial Clerodane Type Diterpenoid from Dodonaea viscosa (L.) Jaeq. Int. J. Mol. Sci. 2015, 16 (9), 20290-20307.