MOPS, 1.0M buffer soln., pH 6.5
MOPS, 1.0M buffer soln., pH 6.5
MOPS, 1.0M buffer soln., pH 6.5
Thermo Scientific Chemicals

MOPS, 1.0M buffer soln., pH 6.5

CAS: 1132-61-2 | C7H15NO4S | 209.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
500 mL
Catalog number J61797.AK
also known as J61797-AK
Price (EUR)
88,65
Online Exclusive
97,90
Save 9,25 (9%)
Each
Quantity:
250 mL
Request bulk or custom format
Price (EUR)
88,65
Online Exclusive
97,90
Save 9,25 (9%)
Each
Specifications
CAS1132-61-2
ChEBICHEBI:44115
Chemical Name or MaterialMOPS buffer soln.
ColorColorless
Health Hazard 1H315-H319-H335
View more
MOPS is frequently used as a buffering agent in biology and biochemistry. It used widely for polyacrylamide gel electrophoresis. It is used as a morpholino propanesulfonic acid buffer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
MOPS is frequently used as a buffering agent in biology and biochemistry. It used widely for polyacrylamide gel electrophoresis. It is used as a morpholino propanesulfonic acid buffer.

Solubility
Soluble in water.

Notes
Usage above 20 mM in mammalian cell culture work is not recommended. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents, Strong bases.
RUO – Research Use Only
MM Santoro.; DW Bolen. A test of the linear extrapolation of unfolding free energy changes over an extended denaturant concentration range. Biochemistry. 1992, 31, (20), 4901-4907.