Alginic acid sodium salt, high viscosity
Alginic acid sodium salt, high viscosity
Alginic acid sodium salt, high viscosity
Alginic acid sodium salt, high viscosity
Thermo Scientific Chemicals

Alginic acid sodium salt, high viscosity

A gelling polysaccharide extracted from giant brown seaweed, useful in viscosity studies | CAS: 9005-38-3 | (C6H7O7)A(C6H7O7)BNa
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
250 g
1 kg
Catalog number J61887.22
also known as J61887-22
Price (EUR)
61,65
キャンペーン価格
68,10
Save 6,45 (9%)
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
61,65
キャンペーン価格
68,10
Save 6,45 (9%)
Each
Chemical Identifiers
CAS9005-38-3
Molecular Formula(C6H7O7)A(C6H7O7)BNa
SMILES[Na+].O[C@H]1[C@@H](O)[C@H](O-*)[C@H](O[C@H]1O-*)C([O-])=O.O[C@H]1[C@@H](O)[C@H](O-*)[C@@H](O[C@H]1O-*)C([O-])=O
Molecular Weight (g/mol)NaN
SpecificationsSpecification SheetSpecification Sheet
Assay (special)Lead: ≤5 ppm
Loss on Drying≤15.0%
Viscosity1000 to 1500 mPa's (1% solution)
FormPowder
Assay (special)Arsenic: ≤3 ppm
View more
A gelling polysaccharide extracted from giant brown seaweed, useful in viscosity studiesAlginic acid sodium salt is widely utilized in food industry as flavorless gum, which enhances viscosity and as an emulsifier. It finds application as thickener for reactive dyestuffs, which is used in textile screen printing and carpet jet-printing. It is used indigestion tablets and the preparation of dental impressions. As a polymer, it combines with chitosan to synthesize a biodegradable porous scaffold, which is useful for tissue engineering applications. It acts as a food additive and involved in the production of gel-like foods such as bakers. It is employed as thickener for reactive dyestuffs in textile screen-printing and carpet jet-printing. It is also used as encapsulating agents of microparticles of beta-galactosidae and in the preparation of alginate hydrogels with tunable mechanical properties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A gelling polysaccharide extracted from giant brown seaweed, useful in viscosity studiesAlginic acid sodium salt is widely utilized in food industry as flavorless gum, which enhances viscosity and as an emulsifier. It finds application as thickener for reactive dyestuffs, which is used in textile screen printing and carpet jet-printing. It is used indigestion tablets and the preparation of dental impressions. As a polymer, it combines with chitosan to synthesize a biodegradable porous scaffold, which is useful for tissue engineering applications. It acts as a food additive and involved in the production of gel-like foods such as bakers. It is employed as thickener for reactive dyestuffs in textile screen-printing and carpet jet-printing. It is also used as encapsulating agents of microparticles of beta-galactosidae and in the preparation of alginate hydrogels with tunable mechanical properties.

Solubility
Soluble in water. Insoluble in alcohol, chloroform and ether.

Notes
Incompatible with strong oxidizing agents, acids and bases.
RUO – Research Use Only

General References:

  1. Vicini, S.; Castellano, M.; Mauri, M.; Marsano, E. Gelling process for sodium alginate: New technical approach by using calcium rich micro-spheres Carbohydr Polym. 2015, 134, 764-774.
  2. Levic, S.; Lijaković, I. P.; Dordevic, V.; Rac, V.; Rakic, V.; Knudsen, T. S.; Pavlovic, V.; Bugarski, B.; Nedovic, V. Characterization of sodium alginate/D-limonene emulsions and respective calcium alginate/D-limonene beads produced by electrostatic extrusion. Food Hydrocolloids. 2015, 45, 111-123.

Frequently asked questions (FAQs)

What is the molecular weight of Alginic Acid sodium salt, high viscosity (Cat. No. J61887.xx)?

Based on the viscosity of the chemical (1335 mPa= 1335 cps), the approximate molecular weight of Alginic Acid sodium salt, high viscosity is 120,000 Da.