Dextran sulfate sodium salt, MW ca 8,000
Dextran sulfate sodium salt, MW ca 8,000
Dextran sulfate sodium salt, MW ca 8,000
Thermo Scientific Chemicals

Dextran sulfate sodium salt, MW ca 8,000

CAS: 9011-18-1 | (C6H7O5)m(C6H7O5)n
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
1 kg
Catalog number J62101.14
also known as J62101-14
Price (EUR)
76,30
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
76,30
Each
Chemical Identifiers
CAS9011-18-1
IUPAC NameDextran sulfate sodium salt
MDL NumberMFCD00081551
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale brown
FormPowder
Assay (unspecified)Glucose: 35 to 48%
Assay (unspecified)Specific rotation: +75? to +105?
Assay (unspecified)Specific viscosity: 0.018 to 0.032
View more
Dextran sulfate is used asa nucleic acid hybridization accelerant. The ability of the polymer to precipitate fibrinogen and low-density lipoproteins from plasma solutions has been used for analysis and for anticoagulant applications. Dextran Sulfate Sodium (DSS) which has proven to have versatile value in Biomedical/clinical research. DSS can be used to study Colitis (Inflammatory Bowel disease) as well as an antiviral as agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only