beta-Casomorphin, human, 95+%
beta-Casomorphin, human, 95+%
beta-Casomorphin, human, 95+%
Thermo Scientific Chemicals

beta-Casomorphin, human, 95+%

An opioid peptide
Have Questions?
Change viewbuttonViewtableView
Quantity:
5mg
25mg
Catalog number J62271.LB0
also known as J62271-#0
Price (EUR)
51,70
Each
Quantity:
5mg
Request bulk or custom format
Price (EUR)
51,70
Each
Specifications
CAS102029-74-3
ColorWhite
FormatPowder
Molecular Weight (g/mol)864.01
Product Typebeta-Casomorphin
View more
An opioid peptide

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only
Contents & Storage
Store at -20°C