Erythromycin, Cell Culture Grade
Erythromycin, Cell Culture Grade
Erythromycin, Cell Culture Grade
Thermo Scientific™

Erythromycin, Cell Culture Grade

Erythromycin (E-Mycin or erythrocin), CAS # 114-07-8, is a broad-spectrum antibiotic. | CAS: 114-07-8 | C37H67NO13 | 733.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J62279.14
also known as J62279-14
Price (EUR)
194,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
194,00
Each
Chemical Identifiers
CAS114-07-8
IUPAC Name(3R,4S,5S,6R,7R,9R,11R,12R,13S,14R)-6-{[(2S,3R,4S,6R)-4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl]oxy}-14-ethyl-7,12,13-trihydroxy-4-{[(2R,4R,5S,6S)-5-hydroxy-4-methoxy-4,6-dimethyloxan-2-yl]oxy}-3,5,7,9,11,13-hexamethyl-1-oxacyclotetradecane-2,10-dione
Molecular FormulaC37H67NO13
InChI KeyULGZDMOVFRHVEP-RWJQBGPGSA-N
SMILESCC[C@H]1OC(=O)[C@H](C)[C@@H](O[C@H]2C[C@@](C)(OC)[C@@H](O)[C@H](C)O2)[C@H](C)[C@@H](O[C@@H]2O[C@H](C)C[C@@H]([C@H]2O)N(C)C)[C@](C)(O)C[C@@H](C)C(=O)[C@H](C)[C@@H](O)[C@]1(C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA93.0-100.5% (sum of erythromycin A, B and C, dry weight basis)
Residue on ignition≤0.2%
Optical Rotation-74.5° ±3.5°
FormCrystals or powder or crystalline powder
Residual water≤6.5%
View more
Erythromycin, is used as a macrolide antibiotic protein synthesis inhibitor. It is widely used in cell culture applications. Depending upon the strain of bacteria, erythromycin has been used between 50 and 200 mg/L to control bacterial growth. Erythromycin resistance can be induced in bacteria.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Erythromycin is a macrolide antibiotic that works by binding the 50S ribosomal subunit and inhibiting polypeptide formation
  • Erythromycin was isolated from Saccharopolyspora erythraea

Applications

  • Erythromycin can be used to disrupt the growth of gram-positive and gram-negative bacteria
  • Erythromycin has been shown to have motor effects in the gastrointestinal system and can be used for laboratory research on migrating motor complex activity
RUO – Research Use Only

General References:

  1. Dreno, B.; Martin, R.; Moyal, D.; Henley, J.B.; Khammari, A.; Seité, S. Skin microbiome and acne vulgaris: Staphylococcus, a new actor in acne. Exp Dermatol. 2017, 26(9), 798-803.
  2. Wu, F.; Tokach, M.D.; DeRouchey, J.M.; Dritz, S.S.; Woodworth, J.C.; Goodband, R.D.; Chitakasempornkul, K.; Bello, N.M.; Capps, K.; Remfry, S.; Scott, H.M.; Nagaraja, T.G.; Apley, M.D.; Amachawadi, R.G. Effects of Tylosin Administration Routes on the Prevalence of Antimicrobial Resistance Among Fecal Enterococci of Finishing Swine. Foodborne Pathog Dis. 2019, 16(5), 309-316.