Bumetanide, 98+%
Bumetanide, 98+%
Bumetanide, 98+%
Thermo Scientific Chemicals

Bumetanide, 98+%

Inhibitor of the sodium-potassium-chloride cotransporter | CAS: 28395-03-1 | C17H20N2O5S | 364.416 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J62302.03
also known as J62302-03
Price (EUR)
122,65
キャンペーン価格
136,00
Save 13,35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
122,65
キャンペーン価格
136,00
Save 13,35 (10%)
Each
Specifications
CAS28395-03-1
ChEBICHEBI:3213
Chemical Name or MaterialBumetanide
ColorWhite
EINECS Number249-004-6
View more
Bumetanide has been observed to block the depolarization of GABA in cortical development studies. It is been used as a specific inhibitor of Na+-K+-2Cl- cotransporter NKCC1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bumetanide has been observed to block the depolarization of GABA in cortical development studies. It is been used as a specific inhibitor of Na+-K+-2Cl- cotransporter NKCC1

Solubility
Soluble in ethanol (10 mg/ml), DMSO (25 mg/ml), acetone, benzene, methanol, propylene glycol, and water (<1mg/mL).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from acids.
RUO – Research Use Only

General References:

  1. Star, R.A., et al. Luminal disequilibrium pH and ammonia transport in outer medullary collecting duct. Am. J. Physiol. 1987, 252( 6 Pt 2), F1148-F1157.
  2. O'Grady, S.M., et al. Characteristics and functions of Na-K-Cl cotransport in epithelial tissues. Am. J. Physiol. 1987, 253( 2 Pt 1), C177-C192.