Thermo Scientific Chemicals

AMPSO, 98+%

Catalog number: J62378.22
100 g, Each
Thermo Scientific Chemicals

AMPSO, 98+%

Catalog number: J62378.22
100 g, Each
Quantity
Catalog number: J62378.22
also known as J62378-22
Price (EUR)
Price: 142,00
Online price: 127,80
Your price:
Quantity
-

Chemical Identifiers

CAS
68399-79-1
IUPAC Name
2-hydroxy-3-[(1-hydroxy-2-methylpropan-2-yl)amino]propane-1-sulfonic acid
Molecular Formula
C7H17NO5S
InChI Key
ACERFIHBIWMFOR-UHFFFAOYNA-N
SMILES
CC(C)(CO)NCC(O)CS(O)(=O)=O
Loss on Drying
≤ 1.0%
Assay (unspecified)
>98.0%
Identification (FTIR)
Pass
Form
Powder
Solution Test
Pass

Description

It is used as an alkaline transfer buffer used for the transfer of strongly basic proteins from gels to nitrocellulose.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an alkaline transfer buffer used for the transfer of strongly basic proteins from gels to nitrocellulose.

Solubility
Soluble in water (333.33 mg/ml, clear to slightly hazy colorless solution).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text