Glycine, 99+%, Cell Culture Reagent
Glycine, 99+%, Cell Culture Reagent
Glycine, 99+%, Cell Culture Reagent
Thermo Scientific Chemicals

Glycine, 99+%, Cell Culture Reagent

CAS: 56-40-6 | C2H5NO2 | 75.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
1 kg
Catalog number J62407.22
also known as J62407-22
Price (EUR)
22,60
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
22,60
Each
Chemical Identifiers
CAS56-40-6
IUPAC Name2-aminoacetic acid
Molecular FormulaC2H5NO2
InChI KeyDHMQDGOQFOQNFH-UHFFFAOYSA-N
SMILESNCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder or crystals or crystalline powder
Assay from Suppliers CofA≥99.0% (unspecified)
Loss on Drying≤0.2%
This buffer solution is used for eluting (dissociating) antibodies in various antibody purification procedures such as affinity purification of antibody using peptide coupled to a column. It is widely used as a buffer for a variety of immunological applications. Glycine is also used as an eluent in column chromatography. Glycine has been reported for use in the enzymatic determination of lactate using lactic dehydrogenase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
This buffer solution is used for eluting (dissociating) antibodies in various antibody purification procedures such as affinity purification of antibody using peptide coupled to a column. It is widely used as a buffer for a variety of immunological applications. Glycine is also used as an eluent in column chromatography. Glycine has been reported for use in the enzymatic determination of lactate using lactic dehydrogenase.

Solubility
Soluble in water (100 mg/ml), and acetone (slightly soluble). Insoluble in alcohol, and ether.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Recommended storage temperature 2 - 8°C
RUO – Research Use Only
Y Nagata.; MM Burger. Wheat germ agglutinin molecular characteristics and specificity for sugar binding. The Journal of Biological Chemistry. 1974, 249, (2), 3116-3122.