TRIS-buffered saline (TBS, 10X, high salt) pH 7.4
TRIS-buffered saline (TBS, 10X, high salt) pH 7.4
TRIS-buffered saline (TBS, 10X, high salt) pH 7.4
Thermo Scientific Chemicals

TRIS-buffered saline (TBS, 10X, high salt) pH 7.4

CAS: 77-86-1 | C4H11NO3 | 121.136 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
2 L
Catalog number J62664.K2
also known as J62664-K2
Price (EUR)
71,40
Each
Quantity:
1 L
Request bulk or custom format
Price (EUR)
71,40
Each
Specifications
CAS77-86-1
ChEBICHEBI:9754
Chemical Name or MaterialTRIS-buffered saline
IUPAC Name2-amino-2-(hydroxymethyl)propane-1,3-diol
InChI KeyLENZDBCJOHFCAS-UHFFFAOYSA-N
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only