Dextran sulfate sodium salt, MW ca >500,000
Dextran sulfate sodium salt, MW ca >500,000
Dextran sulfate sodium salt, MW ca >500,000
Thermo Scientific Chemicals

Dextran sulfate sodium salt, MW ca >500,000

CAS: 9011-18-1 | (C6H7O5)m(C6H7O5)n | 40,000 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
50 g
100 g
1 kg
Catalog number J62787.22
also known as J62787-22
Price (EUR)
227,65
Online exclusive
253,00
Save 25,35 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
227,65
Online exclusive
253,00
Save 25,35 (10%)
Each
Chemical Identifiers
CAS9011-18-1
IUPAC NameDextran sulfate sodium salt
MDL NumberMFCD00081551
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale yellow
FormPowder
Assay (unspecified)Specific viscosity: 0.10 to 0.70
Assay (special)Total Sulfur content: 17.0 to 20.0%
Chloride content≤1000 ppm
View more
Dextran sulfate sodium is used to study colitis (inflammatory bowel disease) as well as an antiviral agent. It functions as a nonsurfactant, suspending agent with gel forming and viscosity controlling properties. It can be used in gels and moisturizers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only