Agarose Gel Loading Dye (6X, Ficoll-Based)
Thermo Scientific Chemicals

Agarose Gel Loading Dye (6X, Ficoll-Based)

Have Questions?
Catalog NumberQuantity
J62800.AC
also known as J62800-AC
25 mL
Catalog number J62800.AC
also known as J62800-AC
Price (EUR)
94,60
Each
Quantity:
25 mL
Request bulk or custom format
Price (EUR)
94,60
Each
Specifications
Chemical Name or MaterialAgarose gel loading dye
Concentration6X
MDL NumberMFCD00675058
Name NoteFicoll based
Recommended StorageKeep cold
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only