Benzamidine hydrochloride hydrate, 99%, water ca 10-14%
Benzamidine hydrochloride hydrate, 99%, water ca 10-14%
Benzamidine hydrochloride hydrate, 99%, water ca 10-14%
Thermo Scientific Chemicals

Benzamidine hydrochloride hydrate, 99%, water ca 10-14%

Reversible inhibitor of trypsin, trypsin-like proteins and serine proteases | CAS: 206752-36-5 | C7H9ClN2 | 156.61 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
10 g
Catalog number J62823.06
also known as J62823-06
Price (EUR)
39,50
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
39,50
Each
Chemical Identifiers
CAS206752-36-5
IUPAC Namebenzenecarboximidamide hydrochloride
Molecular FormulaC7H9ClN2
InChI KeyLZCZIHQBSCVGRD-UHFFFAOYSA-N
SMILESCl.NC(=N)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥98.5%
Appearance (Color)White
Water Content (Karl Fischer Titration)10.0-14.0%
FormPowder
Reversible inhibitor of trypsin, trypsin-like proteins and serine proteases

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only