Chromomycin A3, 97%
Chromomycin A3, 97%
Chromomycin A3, 97%
Thermo Scientific Chemicals

Chromomycin A3, 97%

CAS: 7059-24-7 | C57H82O26 | 1183.257 g/mol
Have Questions?
Catalog NumberQuantity
J62927.MA
also known as J62927-MA
10 mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J62927.MA
also known as J62927-MA
Price (EUR)
391,00
Each
Quantity:
10 mg
Request bulk or custom format
Price (EUR)
391,00
Each
Chemical Identifiers
CAS7059-24-7
IUPAC Name(2R,3S,4R,6S)-6-{[(6S,7S)-6-{[(2S,4R,5R,6R)-4-{[(2S,4R,5R,6R)-4-{[(2S,4S,5S,6S)-5-(acetyloxy)-4-hydroxy-4,6-dimethyloxan-2-yl]oxy}-5-hydroxy-6-methyloxan-2-yl]oxy}-5-hydroxy-6-methyloxan-2-yl]oxy}-7-[(1S,3S,4R)-3,4-dihydroxy-1-methoxy-2-oxopentyl]-4,10-dihydroxy-3-methyl-5-oxo-5,6,7,8-tetrahydroanthracen-2-yl]oxy}-4-{[(2R,4R,5R,6R)-4-hydroxy-5-methoxy-6-methyloxan-2-yl]oxy}-2-methyloxan-3-yl acetate
Molecular FormulaC57H82O26
InChI KeyZYVSOIYQKUDENJ-WKSBCEQHSA-N
SMILESCO[C@@H]([C@@H]1CC2=CC3=CC(O[C@H]4C[C@@H](O[C@@H]5C[C@@H](O)[C@@H](OC)[C@@H](C)O5)[C@@H](OC(C)=O)[C@@H](C)O4)=C(C)C(O)=C3C(O)=C2C(=O)[C@H]1O[C@H]1C[C@@H](O[C@H]2C[C@@H](O[C@H]3C[C@](C)(O)[C@@H](OC(C)=O)[C@H](C)O3)[C@H](O)[C@@H](C)O2)[C@H](O)[C@@H](C)O1)C(=O)[C@@H](O)[C@@H](C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
FormPowder or crystals or crystalline powder
Assay from Supplier's CofA≥96.0%
Chromomycin A3, is used as a G-C specific DNA ligand which inhibits transcription and acts as a DNA binding dye. Chromomycin A3 antagonizes enhanced DNA binding of the transcription factors Sp1 and Sp3 to their cognate G-C box, induced by oxidative stress or DNA damage. Furthermore, by inhibiting transcription, Chromomycin A3 and its structural analogs can inhibit protein biosynthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chromomycin A3, is used as a G-C specific DNA ligand which inhibits transcription and acts as a DNA binding dye. Chromomycin A3 antagonizes enhanced DNA binding of the transcription factors Sp1 and Sp3 to their cognate ″G-C&Pr
RUO – Research Use Only

General References:

  1. CT Amemiya.;JR Gold. Chromomycin A3 stains nucleolus organizer regions of fish chromosomes. Copeia. 19861986 (1), 226-231.
  2. RH Jensen. Chromomycin A3 as a fluorescent probe for flow cytometry of human gynecologic samples. Journal of Histochemistry & Cytochemistry. 197725 (7), 573-579.