E-64
E-64
E-64
Thermo Scientific Chemicals

E-64

Selective and irreversible cysteine protease inhibitor | CAS: 66701-25-5 | C15H27N5O5 | 357.41 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mg
25 mg
Catalog number J62933.LB0
also known as J62933-#0
Price (EUR)
116,00
Each
Quantity:
5 mg
Request bulk or custom format
Price (EUR)
116,00
Each
Chemical Identifiers
CAS66701-25-5
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder
Assay from Supplier's CofA≥98.0% (HPLC)
It is used as a broad spectrum cysteine proteinase and calpain activation inhibitor. Additionally, Prevotella intermediate studies report that E-64 blocks the breakdown of methaemoglobin and the oxidation of oxyhaemoglobin by InpA. E-64 has also been used to study excystation in Giardia lamblia. E-64 is an inhibitor of Calpain, cathepsin K and cathepsin S. E-64 is an effective ligand for affinity purification of cysteine proteases. When coupled to a thiolated affinity matrix, binding is no longer irreversible, but specificity is retained

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a broad spectrum cysteine proteinase and calpain activation inhibitor. Additionally, Prevotella intermediate studies report that E-64 blocks the breakdown of methaemoglobin and the oxidation of oxyhaemoglobin by InpA. E-64 has also been used to study excystation in Giardia lamblia. E-64 is an inhibitor of Calpain, cathepsin K and cathepsin S. E-64 is an effective ligand for affinity purification of cysteine proteases. When coupled to a thiolated affinity matrix, binding is no longer irreversible, but specificity is retained

Solubility
Soluble in DMSO (25mg/ml), or water (20mg/ml)

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C. Keep away from strong acids, strong bases.
RUO – Research Use Only

General References:

  1. Susa et al. Human primary osteoclasts: in vitro generation and applications as pharmacological and clinical assay. J.Transl.Med. 2004, 2, (1), 6-9.
  2. Zhao et al. Cytosolic FoxO1 is essential for the induction of autophagy and tumour suppressor activity. Nat.Cell.Biol. 2010, 12, (7), 665-75.