Trazodone hydrochloride, 98+%
Trazodone hydrochloride, 98+%
Trazodone hydrochloride, 98+%
Thermo Scientific Chemicals

Trazodone hydrochloride, 98+%

A serotonin uptake inhibitor | CAS: 25332-39-2 | C19H23Cl2N5O | 408.327 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J63070.03
also known as J63070-03
Price (EUR)
78,50
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
78,50
Each
Chemical Identifiers
CAS25332-39-2
IUPAC Namehydrogen 2-{3-[4-(3-chlorophenyl)piperazin-1-yl]propyl}-2H,3H-[1,2,4]triazolo[4,3-a]pyridin-3-one chloride
Molecular FormulaC19H23Cl2N5O
InChI KeyOHHDIOKRWWOXMT-UHFFFAOYSA-N
SMILES[H+].[Cl-].ClC1=CC=CC(=C1)N1CCN(CCCN2N=C3C=CC=CN3C2=O)CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale yellow
FormPowder or crystals or crystalline powder
Assay (unspecified)≥98.0%
A serotonin uptake inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only