L-Cystine disodium salt monohydrate, 98+%
L-Cystine disodium salt monohydrate, 98+%
L-Cystine disodium salt monohydrate, 98+%
Thermo Scientific Chemicals

L-Cystine disodium salt monohydrate, 98+%

CAS: 199329-53-8 | C6H12N2Na2O5S2 | 302.271 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number J63071.18
also known as J63071-18
Price (EUR)
66,10
Each
Quantity:
50 g
Request bulk or custom format
Price (EUR)
66,10
Each
Chemical Identifiers
CAS199329-53-8
IUPAC Namedisodium 2-amino-3-[(2-amino-2-carboxylatoethyl)disulfanyl]propanoate hydrate
Molecular FormulaC6H12N2Na2O5S2
InChI KeyTVPDKJXPEMKMDM-UHFFFAOYNA-L
SMILESO.[Na+].[Na+].NC(CSSCC(N)C([O-])=O)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale yellow
FormPowder
Assay from Supplier's CofA≥98.0% (Titration)
Water Content (Karl Fischer Titration)≤8.0%
Solution TestClear and complete (1.0% solution in Water)
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only