Neuraminidase, Clostridium perfringens
Thermo Scientific Chemicals

Neuraminidase, Clostridium perfringens

CAS: 9001-67-6
Have Questions?
Catalog NumberQuantityPackaging
J63259.MA
also known as J63259-MA
10 mgPlastic bottle
Catalog number J63259.MA
also known as J63259-MA
Price (EUR)
305,65
Online Exclusive
339,00
Save 33,35 (10%)
Each
Quantity:
10 mg
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
305,65
Online Exclusive
339,00
Save 33,35 (10%)
Each
Chemical Identifiers
CAS9001-67-6
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)A280nm: As reported
Unit definitionOne unit liberates one ?mole of sialic acid per minute at 37?C, pH 5.0 from bovine submaxillary mucin
units/mgP0.5 units/mg min.
Neuraminidase is used as a hydrolytic enzyme that removes sialic acid from mucoproteins, used in a study to assess binding with human T lymphocytes in sheep pretreated with neuraminidase. It has also been used in a study to investigate the effect of bile salts on the action of hydrolysis by neuraminidase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only