Honokiol, 98+%
Honokiol, 98+%
Honokiol, 98+%
Thermo Scientific Chemicals

Honokiol, 98+%

Induces apoptosis in human squamous lung cancer cells | CAS: 35354-74-6 | C18H18O2 | 266.34 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mg
25 mg
100 mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J63434.MA
also known as J63434-MA
Price (EUR)
116,00
Each
Quantity:
10 mg
Request bulk or custom format
Price (EUR)
116,00
Each
Specifications
CAS35354-74-6
ChEBICHEBI:5759
Chemical Name or MaterialHonokiol
ColorWhite
DOT InformationTransport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
View more
Induces apoptosis in human squamous lung cancer cells

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only