1-Aminobenzotriazole, 97%
1-Aminobenzotriazole, 97%
1-Aminobenzotriazole, 97%
Thermo Scientific Chemicals

1-Aminobenzotriazole, 97%

Suicide inhibitor of cytochrome P450 and chloroperoxidase | CAS: 1614-12-6 | C6H6N4 | 134.142 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number J63610.06
also known as J63610-06
Price (EUR)
1.049,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
1.049,00
Each
Specifications
Beilstein607843
CAS1614-12-6
Chemical Name or Material1-Aminobenzotriazole
ColorBrown to White
Formula Weight134.14
View more
1-Aminobenzotriazole is used as a Suicide inhibitor of cytochrome P450 and chloroperoxidase, an inhibitor of oxidative drug metabolism in biochemical pharmacology and toxicology studies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Aminobenzotriazole is used as a Suicide inhibitor of cytochrome P450 and chloroperoxidase, an inhibitor of oxidative drug metabolism in biochemical pharmacology and toxicology studies.

Solubility
Soluble in chloroform, DMSO, ethanol or water

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
RUO – Research Use Only

General References:

  1. Elizabeth A. Dierks.; Christopher Davis, S.; Paul R. Ortiz de Montellano. Glu-320 and Asp-323 Are Determinants of the CYP4A1 Hydroxylation Regiospecificity and Resistance to Inactivation by 1-Aminobenzotriazole.Biochemistry.1998, (7), 1839-1847.
  2. Miranda, C.L.; Henderson, M.C.; Buhler, D.R. Evaluation of Chemicals as Inhibitors of Trout Cytochrome P450s.Toxicology and Applied Pharmacology.1998, (2), 237-244.