CHES, 0.5M buffer soln, pH 9.0
CHES, 0.5M buffer soln, pH 9.0
CHES, 0.5M buffer soln, pH 9.0
CHES, 0.5M buffer soln, pH 9.0
Thermo Scientific Chemicals

CHES, 0.5M buffer soln, pH 9.0

CAS: 103-47-9 | C8H17NO3S | 207.288 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
250 mL
Catalog number J63670.AK
also known as J63670-AK
Price (EUR)
102,85
Online Exclusive
121,00
Save 18,15 (15%)
Each
Add to cart
Quantity:
250 mL
Request bulk or custom format
Price (EUR)
102,85
Online Exclusive
121,00
Save 18,15 (15%)
Each
Add to cart
Specifications
CAS103-47-9
Chemical Name or MaterialCHES buffer soln.
Health Hazard 1Warning
Health Hazard 2GHS H Statement
H319
Causes serious eye irritation.
Health Hazard 3GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
If eye irritation persists: Get medical advice/attention.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only