Nimesulide
Nimesulide
Nimesulide
Thermo Scientific Chemicals

Nimesulide

Highly selective cyclooxygenase-2 inhibitor which induces apoptosis | CAS: 51803-78-2 | C13H12N2O5S | 308.308 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J63699.06
also known as J63699-06
Price (EUR)
60,20
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
60,20
Each
Chemical Identifiers
CAS51803-78-2
SpecificationsSpecification SheetSpecification Sheet
Assay≥97.0%
Assay (unspecified)≥97.0%
Nimesulide are used for the treatment of acute pain, the symptomatic treatment of osteoarthritis and primary dysmenorrhoea, . Nimesulide was as effective as paracetamol in reducing fever, local pain, and general discomfort. It was effective in reducing pain associated with osteoarthritis, cancer, thrombophlebitis, oral surgery in adults, otorhinolaryngological diseases and traumatic injury. Nimesulide is demonstrated to suppress azoxymethane-induced carcinogenesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water (<50 µg/ml), 1:10 DMSO:PBS (pH 7.2) (<200 µg/ml), ethanol (1 mg/ml), DMSO (15 mg/ml), DMF (15 mg/ml), chloroform, dichloromethane, acetone (freely soluble), and 1N NaOH.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Rick Davis,; Rex N. Brogden.Nimesulide, An Update of its Pharmacodynamic and Pharmacokinetic Properties, and Therapeutic Efficacy. Drugs. Drugs, 199448 (3), 431-454.
  2. Alberto Bernaregg. Clinical Pharmacokinetics of Nimesulide. Clinical Pharmacokinetics. 1995, 35 (4), 247-274.