Brompheniramine maleate
Brompheniramine maleate
Brompheniramine maleate
Thermo Scientific Chemicals

Brompheniramine maleate

An antihistamine and also an inhibitor of serotonin reuptake | CAS: 980-71-2 | C20H23BrN2O4 | 435.32 g/mol
Have Questions?
Catalog NumberQuantity
J63922.MB
also known as J63922-MB
25 mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J63922.MB
also known as J63922-MB
Price (EUR)
421,00
Each
Quantity:
25 mg
Request bulk or custom format
Price (EUR)
421,00
Each
Chemical Identifiers
CAS980-71-2
IUPAC Name(2Z)-but-2-enedioic acid; [3-(4-bromophenyl)-3-(pyridin-2-yl)propyl]dimethylamine
Molecular FormulaC20H23BrN2O4
InChI KeySRGKFVAASLQVBO-BTJKTKAUNA-N
SMILESOC(=O)\C=C/C(O)=O.CN(C)CCC(C1=CC=C(Br)C=C1)C1=CC=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
FormWhite to off-white crystalline powder
Melting Point136 to 138? C
Assay (HPLC)≥ 99%
An antihistamine and also an inhibitor of serotonin reuptake

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only