Salubrinal
Salubrinal
Salubrinal
Thermo Scientific Chemicals

Salubrinal

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mg
10 mg
Catalog number J64192.MA
also known as J64192-MA
Price (EUR)
190,65
Online Exclusive
212,00
Save 21,35 (10%)
Each
Quantity:
10 mg
Request bulk or custom format
Price (EUR)
190,65
Online Exclusive
212,00
Save 21,35 (10%)
Each
Chemical Identifiers
CAS405060-95-9
IUPAC Name3-phenyl-N-(2,2,2-trichloro-1-{[(quinolin-8-yl)carbamothioyl]amino}ethyl)prop-2-enamide
Molecular FormulaC21H17Cl3N4OS
InChI KeyLCOIAYJMPKXARU-UHFFFAOYNA-N
SMILESClC(Cl)(Cl)C(NC(=S)NC1=C2N=CC=CC2=CC=C1)NC(=O)C=CC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)Cream to brown
Purity≥95.0%
Salubrinal is a selective Inhibitor of eIF2α (eukaryotic translation initiation factor 2 α-subunit) dephosphorylation, inhibiting global protein translation. Salubrinal is a specific inhibitor of ER Stress induced apoptosis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only
Contents & Storage
2°C to 8°C