Pifithrin-alpha, p-Nitro hydrobromide, Thermo Scientific Chemicals
Pifithrin-alpha, p-Nitro hydrobromide, Thermo Scientific Chemicals
Pifithrin-alpha, p-Nitro hydrobromide, Thermo Scientific Chemicals
Pifithrin-alpha, p-Nitro hydrobromide, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Pifithrin-alpha, p-Nitro hydrobromide, Thermo Scientific Chemicals

 
Have Questions?
Catalog NumberQuantity
J64277.LB0
also known as J64277-#0
5 mg
Catalog number J64277.LB0
also known as J64277-#0
Price (EUR)
96,80
Each
Add to cart
Quantity:
5 mg
Request bulk or custom format
Price (EUR)
96,80
Each
Add to cart
Chemical Identifiers
CAS389850-21-9
IUPAC Name2-(2-imino-2,3,4,5,6,7-hexahydro-1,3-benzothiazol-3-yl)-1-(4-nitrophenyl)ethan-1-one hydrobromide
Molecular FormulaC15H16BrN3O3S
InChI KeyNXPCMOGORSWOLH-UHFFFAOYSA-N
SMILESBr.[O-][N+](=O)C1=CC=C(C=C1)C(=O)CN1C(=N)SC2=C1CCCC2
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Cream
Purity≥97.0%
FormSolid
A cell-permeable p53 inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only