2'-Amino-3'-methoxyflavone, 99%
2'-Amino-3'-methoxyflavone, 99%
2'-Amino-3'-methoxyflavone, 99%
Thermo Scientific Chemicals

2'-Amino-3'-methoxyflavone, 99%

A MAP kinase inhibitor | CAS: 167869-21-8 | C16H13NO3 | 267.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mg
25 mg
Catalog number J64485.MB
also known as J64485-MB
Price (EUR)
323,00
Each
Quantity:
25 mg
Request bulk or custom format
Price (EUR)
323,00
Each
Specifications
CAS167869-21-8
ChEBICHEBI:77954
Chemical Name or Material2'-Amino-3'-methoxyflavone
ColorYellow
Formula Weight267.28
View more
A MAP kinase inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only