Azadibenzocyclooctyne-PEG4-alcohol
Azadibenzocyclooctyne-PEG4-alcohol
Azadibenzocyclooctyne-PEG4-alcohol
Thermo Scientific Chemicals

Azadibenzocyclooctyne-PEG4-alcohol

ADIBO-containing reagent with PEG4 spacer and alcohol terminus | C29H36N2O7
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mg
100 mg
Catalog number J64617.MB
also known as J64617-MB
Price (EUR)
129,00
Each
Quantity:
25 mg
Request bulk or custom format
Price (EUR)
129,00
Each
Specifications
Chemical Name or MaterialAzadibenzocyclooctyne-PEG4-alcohol
ColorYellow
Formula Weight524.61
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
ADIBO-containing reagent with PEG4 spacer and alcohol terminus

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only