Gly-Phe beta-naphthylamide
Gly-Phe beta-naphthylamide
Gly-Phe beta-naphthylamide
Thermo Scientific Chemicals

Gly-Phe beta-naphthylamide

347.41 g/mol
Have Questions?
Catalog NumberQuantity
J64718.MC
also known as J64718-MC
100 mg
Catalog number J64718.MC
also known as J64718-MC
Price (EUR)
75,65
Online exclusive
83,80
Save 8,15 (10%)
Each
Quantity:
100 mg
Request bulk or custom format
Price (EUR)
75,65
Online exclusive
83,80
Save 8,15 (10%)
Each
Specifications
Common NameGlycylphenylalanine 2-naphthylamide
For Use With (Application)Substrate for cathepsin C
FormatCrystalline
Molecular Weight (g/mol)347.41
Product TypeGly-Phe beta-naphthylamide
View more
Gly-Phe beta-naphthylamide is used to accumulate inside lysosomes leading to their destruction. Studies report that Gly-Phe-β-naphthylamide significantly inhibits the activation of caspase-8, and has also been used to study recombinant cathepsin C.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only
Contents & Storage
-30°C to -10°C