Diphenyleneiodonium chloride
Diphenyleneiodonium chloride
Diphenyleneiodonium chloride
Thermo Scientific Chemicals

Diphenyleneiodonium chloride

CAS: 4673-26-1 | C12H8ClI | 314.55 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mg
100 mg
Catalog number J64838.MC
also known as J64838-MC
Price (EUR)
408,00
Each
Quantity:
100 mg
Request bulk or custom format
Price (EUR)
408,00
Each
Specifications
CAS4673-26-1
ChEBICHEBI:77967
Chemical Name or MaterialDiphenyleneiodonium chloride
ColorWhite
Formula Weight314.55
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only