N-Benzyl-p-toluenesulfonamide
N-Benzyl-p-toluenesulfonamide
N-Benzyl-p-toluenesulfonamide
Thermo Scientific Chemicals

N-Benzyl-p-toluenesulfonamide

A potent inhibitor of calcium-stimulated myosin S1 ATPase | CAS: 1576-37-0 | C14H15NO2S | 261.339 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mg
10 mg
Catalog number J64910.LB0
also known as J64910-#0
Price (EUR)
111,00
Each
Quantity:
5 mg
Request bulk or custom format
Price (EUR)
111,00
Each
Specifications
CAS1576-37-0
Chemical Name or MaterialN-Benzyl-p-toluenesulfonamide
ColorWhite
Formula Weight261.34
Health Hazard 1H315-H319-H335
View more
A potent inhibitor of calcium-stimulated myosin S1 ATPase

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only