N6-Methyl-2'-deoxyadenosine, 99%
N6-Methyl-2'-deoxyadenosine, 99%
N6-Methyl-2'-deoxyadenosine, 99%
Thermo Scientific Chemicals

N6-Methyl-2'-deoxyadenosine, 99%

CAS: 2002-35-9 | C11H15N5O3 | 265.27 g/mol
Have Questions?
Catalog NumberQuantity
J64961.03
also known as J64961-03
1 g
Catalog number J64961.03
also known as J64961-03
Price (EUR)
409,00
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
409,00
Each
Specifications
CAS2002-35-9
ChEBICHEBI:7417
Chemical Name or MaterialN6-Methyl-2'-deoxyadenosine
ColorWhite
Formula Weight265.27
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only