AY 9944 Dihydrochloride, Thermo Scientific Chemicals
AY 9944 Dihydrochloride, Thermo Scientific Chemicals
AY 9944 Dihydrochloride, Thermo Scientific Chemicals
AY 9944 Dihydrochloride, Thermo Scientific Chemicals
Thermo Scientific Chemicals

AY 9944 Dihydrochloride, Thermo Scientific Chemicals

Inhibitor of hedgehog (Hh) signaling
Have Questions?
Catalog NumberQuantity
J65079.MF
also known as J65079-MF
50 mg
Catalog number J65079.MF
also known as J65079-MF
Price (EUR)
777,00
Each
Quantity:
50 mg
Request bulk or custom format
Price (EUR)
777,00
Each
Specifications
CAS366-93-8
DescriptionWhite
For Use With (Application)Inhibitor of hedgehog (Hh) signaling
FormSolid
Formula Weight464.30g/mol
View more
Inhibitor of hedgehog (Hh) signaling

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only
Contents & Storage
2°C to 8°C