Guanidine thiocyanate, Molecular Biology Grade
Guanidine thiocyanate, Molecular Biology Grade
Guanidine thiocyanate, Molecular Biology Grade
Thermo Scientific Chemicals

Guanidine thiocyanate, Molecular Biology Grade

A protein denaturant | CAS: 593-84-0 | CH5N3·CHNS
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
500 g
Catalog number J65104.18
also known as J65104-18
Price (EUR)
76,65
Online exclusive
84,50
Save 7,85 (9%)
Each
Quantity:
50 g
Request bulk or custom format
Price (EUR)
76,65
Online exclusive
84,50
Save 7,85 (9%)
Each
Chemical Identifiers
CAS593-84-0
IUPAC Nameguanidine; sulfanylformonitrile
Molecular FormulaC2H6N4S
InChI KeyZJYYHGLJYGJLLN-UHFFFAOYSA-N
SMILESSC#N.NC(N)=N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white to pale yellow
FormCrystalline powder
Assay≥99.0%
DNaseNone detected
RNaseNone detected
Guanidine thiocyanate is used for RNA isolation and can solubilize proteins. When the compound is used to solubilize proteins it can be removed by dialysis against urea without difficulty. The hybridization of guanidine thiocyanate-solubilize hepatocytes can be used as a method to quantify mRNA from hepatocytes. Blood sample lysate stored in guanidine thiocyanate has been shown to maintain nucleic acid integrity. Research describes guanidine thiocyanate to be ineffective for RNA isolation of plant tissue with a high levels of secondary metabolites and storage proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Guanidine thiocyanate is used for RNA isolation and can solubilize proteins. When the compound is used to solubilize proteins it can be removed by dialysis against urea without difficulty. The hybridization of guanidine thiocyanate-solubilize hepatocytes can be used as a method to quantify mRNA from hepatocytes. Blood sample lysate stored in guanidine thiocyanate has been shown to maintain nucleic acid integrity. Research describes guanidine thiocyanate to be ineffective for RNA isolation of plant tissue with a high levels of secondary metabolites and storage proteins.

Solubility
Soluble in water (1420 g/L) at 20°C, and ethanol (5%).

Notes
Light Sensitive. Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Gregory J. Wadsworth.; Margaret G. Redinbaugh.; John G. Scandalios. A procedure for the small-scale isolation of plant RNA suitable for RNA blot analysis . Analytical Biochemistry. 1988, 172, (1), 279-283.
  2. John M. Chirgwin.; Alan E. Przybyla.; Raymond J. MacDonald.; William J. Rutter. Isolation of biologically active ribonucleic acid from sources enriched in ribonuclease. Biochemistry. 1979, 18, (24), 5294-5299.