5-Bromo-5-nitro-1,3-dioxane, 98%
5-Bromo-5-nitro-1,3-dioxane, 98%
5-Bromo-5-nitro-1,3-dioxane, 98%
Thermo Scientific Chemicals

5-Bromo-5-nitro-1,3-dioxane, 98%

5-Bromo-5-nitro-1,3-dioxane, also known as bronidox, shows antimicrobial activity against gram-negative and gram-positive bacteria, yeast, and fungi. | CAS: 30007-47-7 | C4H6BrNO4 | 211.999 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J65443.14
also known as J65443-14
Price (EUR)
216,65
Online exclusive
241,00
Save 24,35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
216,65
Online exclusive
241,00
Save 24,35 (10%)
Each
Chemical Identifiers
CAS30007-47-7
IUPAC Name5-bromo-5-nitro-1,3-dioxane
Molecular FormulaC4H6BrNO4
InChI KeyXVBRCOKDZVQYAY-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1(Br)COCOC1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)97.5% min.
Melting Pointapprox. 60?C
Proton NMRConforms to structure
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

General Description

• 5-Bromo-5-nitro-1,3-dioxane is a chemical compound showing antimicrobial activity against gram-negative and gram-positive bacteria, yeast, and fungi
• This compound is capable of promoting the oxidation of essential protein thiols, causing inhibition of enzyme activity leading to the inhibition of microbial growth

Application

• 5-Bromo-5-nitro-1,3-dioxane has been used as a stabilizer and preserving agent for biological molecules and solutions, including antibodies and antisera
• It can be used alone or combined with methylisothiazolinone, which is also considered to be an effective preservative

RUO – Research Use Only

General References:

  1. Ya. Legin, G. 2-Bromo-2-nitro-1,3-propanediol(Bronopol) and its derivatives: Synthesis, properties, and application (a review). Pharmaceutical Chemistry Journal. 1996, 30, (4), 273-284.
  2. Ghannoum, M.; Thomson, M.; Bowman, W.; Al-Khalil, S. Mode of action of the antimicrobial compound 5-Bromo-5-nitro-1,3-dioxane (Bronidox). Folia Microbiologica. 1986, 31, (1), 19-31.