Deoxyribonucleic acid sodium salt, salmon testes
Deoxyribonucleic acid sodium salt, salmon testes
Deoxyribonucleic acid sodium salt, salmon testes
Thermo Scientific Chemicals

Deoxyribonucleic acid sodium salt, salmon testes

CAS: 68938-01-2 | C17H15BrFNO3 | 50000-100000 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number J65753.06
also known as J65753-06
Price (EUR)
628,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
628,00
Each
Chemical Identifiers
CAS68938-01-2
IUPAC NameDeoxyribonucleic acid sodium salt
SMILES*
Molecular Weight (g/mol)50000-100000
SynonymDNA sodium salt
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white
FormSolid
Assay (unspecified)E260,1%: 155.0 min.
Assay (unspecified)N:P ratio: 1.5 to 1.8
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only