IGF-1R Inhibitor, PPP
IGF-1R Inhibitor, PPP
IGF-1R Inhibitor, PPP
IGF-1R Inhibitor, PPP
Thermo Scientific Chemicals

IGF-1R Inhibitor, PPP

CAS: 477-47-4 | C22H22O8, C22H22O8 | 414.41 g/mol
Have Questions?
Catalog NumberQuantity
J65916.MA
also known as J65916-MA
10 mg
Catalog number J65916.MA
also known as J65916-MA
Price (EUR)
190,00
Each
Add to cart
Quantity:
10 mg
Request bulk or custom format
Price (EUR)
190,00
Each
Add to cart
Specifications
CAS477-47-4
DescriptionWhite
For Use With (Application)Picropodophyllotoxin is an insulin-like growth factor-I (IGF-I) receptor kinase inhibitor
FormSolid
Formula Weight414.40g/mol
View more
Picropodophyllotoxin is an insulin-like growth factor-I (IGF-I) receptor kinase inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only
Contents & Storage
2°C to 8°C