Leflunomide
Leflunomide
Leflunomide
Thermo Scientific Chemicals

Leflunomide

CAS: 75706-12-6 | C12H9F3N2O2 | 270.211 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
500 mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J65917.MC
also known as J65917-MC
Price (EUR)
137,00
Each
Quantity:
100 mg
Request bulk or custom format
Price (EUR)
137,00
Each
Chemical Identifiers
CAS75706-12-6
IUPAC Name5-methyl-N-[4-(trifluoromethyl)phenyl]-1,2-oxazole-4-carboxamide
Molecular FormulaC12H9F3N2O2
InChI KeyVHOGYURTWQBHIL-UHFFFAOYSA-N
SMILESCC1=C(C=NO1)C(=O)NC1=CC=C(C=C1)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)98.0% min.
An immunosuppressive agent. Inhibits T and B cell proliferation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • An immunosuppressive agent
  • Inhibits T and B cell proliferation
RUO – Research Use Only