2,3,5-Tri-O-benzyl-beta-D-arabinofuranose
2,3,5-Tri-O-benzyl-beta-D-arabinofuranose
2,3,5-Tri-O-benzyl-beta-D-arabinofuranose
2,3,5-Tri-O-benzyl-beta-D-arabinofuranose
Thermo Scientific Chemicals

2,3,5-Tri-O-benzyl-beta-D-arabinofuranose

CAS: 89361-52-4 | C26H28O5 | 420.505 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number J66252.06
also known as J66252-06
Price (EUR)
328,00
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (EUR)
328,00
Each
Add to cart
Specifications
CAS89361-52-4
Chemical Name or Material2,3,5-Tri-O-benzyl-beta-D-ribofuranose
ColorWhite
Formula Weight420.5
IUPAC Name(2R,3R,4R,5R)-3,4-bis(phenylmethoxy)-5-(phenylmethoxymethyl)oxolan-2-ol
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Soluble in chloroform

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
RUO – Research Use Only