Sulfadoxine, 98%
Sulfadoxine, 98%
Sulfadoxine, 98%
Thermo Scientific Chemicals

Sulfadoxine, 98%

CAS: 2447-57-6 | C12H14N4O4S | 310.33 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
View all
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J66268.06
also known as J66268-06
Price (EUR)
48,80
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
48,80
Each
Specifications
Beilstein625453
CAS2447-57-6
ChEBICHEBI:9329
Chemical Name or MaterialSulfadoxine
EINECS Number219-504-9
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only