Sparfloxacin, 98%
Sparfloxacin, 98%
Sparfloxacin, 98%
Thermo Scientific Chemicals

Sparfloxacin, 98%

CAS: 110871-86-8 | C19H22F2N4O3 | 392.41 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
10 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J66358.03
also known as J66358-03
Price (EUR)
38,40
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
38,40
Each
Chemical Identifiers
CAS110871-86-8
IUPAC Name5-amino-1-cyclopropyl-7-[(3R,5S)-3,5-dimethylpiperazin-1-yl]-6,8-difluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic acid
Molecular FormulaC19H22F2N4O3
InChI KeyDZZWHBIBMUVIIW-DTORHVGOSA-N
SMILESC[C@H]1CN(C[C@@H](C)N1)C1=C(F)C(N)=C2C(=O)C(=CN(C3CC3)C2=C1F)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Residue on ignition0.2% max.
Assay (unspecified)98.0% - 102.0%
Impurity contentHeavy metals: 0.002% max.
Loss on Drying1.0% max.
Targets a wide range of Gram-positive, Gram-negative, and mycoplasma species

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only