Sulfamethoxazole, 98%
Sulfamethoxazole, 98%
Sulfamethoxazole, 98%
Thermo Scientific Chemicals

Sulfamethoxazole, 98%

Sulfamethoxazole, CAS # 723-46-6, is a sulfonamide antibiotic that inhibits folic acid synthesis in bacteria and prevents cell division and growth. | CAS: 723-46-6 | C10H11N3O3S | 253.276 g/mol
Have Questions?
Catalog NumberQuantityPackaging
J66565.22
also known as J66565-22
100 gPlastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J66565.22
also known as J66565-22
Price (EUR)
183,65
Online exclusive
204,00
Save 20,35 (10%)
Each
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
183,65
Online exclusive
204,00
Save 20,35 (10%)
Each
Chemical Identifiers
CAS723-46-6
IUPAC Name4-amino-N-(5-methyl-1,2-oxazol-3-yl)benzene-1-sulfonamide
Molecular FormulaC10H11N3O3S
InChI KeyJLKIGFTWXXRPMT-UHFFFAOYSA-N
SMILESCC1=CC(NS(=O)(=O)C2=CC=C(N)C=C2)=NO1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder or crystals or crystalline powder
Loss on Drying≤0.5%
Appearance (Color)White to off-white
Assay (unspecified)≥97.5%
Residue on ignition≤0.5%
View more
A broad spectrum bacteriostatic sulfonamide antibiotic

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Sulfamethoxazole is an antibiotic composed of an isoxazole with a 4-aminobenzenesulfonamido and methyl-group substitution at positions 5 and 3, respectively
  • Sulfamethoxazole carries out most of its action through the inhibition of dihydropteroate synthase, the enzyme responsible for the synthesis of the precursor of folic acid; inhibition of folic acid synthesis prevents DNA replication and cell division in susceptible bacteria

Application

  • Sulfamethoxazole can elicit an allergic response and may be useful for immune cell studies in laboratory settings
  • Sulfamethoxazole is suitable for laboratory research on infection, environmental pollution, and the physiology of susceptible bacteria
RUO – Research Use Only

General References:

  1. Xiong JQ, Govindwar S, Kurade MB, Paeng KJ, Roh HS, Khan MA, Jeon BH. Toxicity of sulfamethazine and sulfamethoxazole and their removal by a green microalga, Scenedesmus obliquus. Chemosphere. 2019 Mar;218:551-558. doi: 10.1016/j.chemosphere.2018.11.146. Epub 2018 Nov 24. PMID: 30500716.
  2. Ma R, Wang Y, Zou X, Hu K, Sun B, Fang W, Fu G, Yang X. Pharmacokinetics of sulfamethoxazole and trimethoprim in Pacific white shrimp, Litopenaeus vannamei, after oral administration of single-dose and multiple-dose. Environ Toxicol Pharmacol. 2017 Jun;52:90-98. doi: 10.1016/j.etap.2017.03.019. Epub 2017 Mar 28. PMID: 28410468.