2-O-alpha-D-Glucopyranosyl-L-ascorbic acid, 98+%
2-O-alpha-D-Glucopyranosyl-L-ascorbic acid, 98+%
2-O-alpha-D-Glucopyranosyl-L-ascorbic acid, 98+%
Thermo Scientific Chemicals

2-O-alpha-D-Glucopyranosyl-L-ascorbic acid, 98+%

CAS: 129499-78-1 | C12H18O11 | 338.265 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number J66601.03
also known as J66601-03
Price (EUR)
32,65
Online exclusive
36,10
Save 3,45 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
32,65
Online exclusive
36,10
Save 3,45 (10%)
Each
Chemical Identifiers
CAS129499-78-1
IUPAC Name(2R)-2-[(1S)-1,2-dihydroxyethyl]-5-hydroxy-4-{[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-2,3-dihydrofuran-3-one
Molecular FormulaC12H18O11
InChI KeyYEERNPPBIMJBHH-DCWMUDTNSA-N
SMILESOC[C@H](O)[C@H]1OC(O)=C(O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)C1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
Assay from Supplier's CofA≥98+% (HPLC)
FormPowder or crystalline powder
Proton NMRConforms to structure
2-O-alpha-D-Glucopyranosyl-L-ascorbic acid reduce the free radicals that result from UV irradiation of the skin, and significantly reduce cell damage and photo-aging. It also promotes collagen synthesis. It can function as an antioxidant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-O-alpha-D-Glucopyranosyl-L-ascorbic acid reduce the free radicals that result from UV irradiation of the skin, and significantly reduce cell damage and photo-aging. It also promotes collagen synthesis. It can function as an antioxidant.

Solubility
Soluble in water. (879 g/L) at 25°C.

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
RUO – Research Use Only

General References:

  1. Itaru Yamamoto. [Course in development of a stable vitamin C derivative (AA-2G)]. Folia Pharmacologica Japonica.2008, 132, (3), 160-165.
  2. Ruizhi Han; Long Liu; Jianghua Li; Guocheng Du; Jian Chen. Functions, applications and production of 2-O-D-glucopyranosyl-L-ascorbic acid. Applied Microbiology and Biotechnology.2012, 95, (2), 313-320.