Sulfamonomethoxine, 98%
Sulfamonomethoxine, 98%
Sulfamonomethoxine, 98%
Thermo Scientific Chemicals

Sulfamonomethoxine, 98%

Long-acting sulfonamide antibacterial agent. It is used to study degradation by Fe3O4 magnetic nanoparticles. It is used in blood kinetic studies and is used to study capsule formation of Bordetella bronchiseptica. | CAS: 1220-83-3 | C11H12N4O3S | 280.302 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J66733.MD
also known as J66733-MD
Price (EUR)
121,00
Each
Quantity:
250 mg
Request bulk or custom format
Price (EUR)
121,00
Each
Specifications
Beilstein621128
CAS1220-83-3
ChEBICHEBI:32164
Chemical Name or MaterialSulfamonomethoxine
Flash Point>110°C (230°F)
View more
Sulfamonomethoxine is a long acting sulfonamide antibacterial agent. It is used to study degradation by Fe3O4 magnetic nanoparticles. It is used in blood kinetic studies and is used to study capsule formation of Bordetella bronchiseptica.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only