Kanamycin disulfate, 700μg/mg, Thermo Scientific Chemicals
Kanamycin disulfate, 700μg/mg, Thermo Scientific Chemicals
Kanamycin disulfate, 700μg/mg, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Kanamycin disulfate, 700μg/mg, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
J66856.06
also known as J66856-06
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J66856.06
also known as J66856-06
Price (EUR)
88,90
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
88,90
Each
Chemical Identifiers
CAS64013-70-3
IUPAC Name2-(aminomethyl)-6-[(4,6-diamino-3-{[4-amino-3,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-2-hydroxycyclohexyl)oxy]oxane-3,4,5-triol; bis(sulfuric acid)
Molecular FormulaC18H40N4O19S2
InChI KeyOGTKIXVMLDAMNU-UHFFFAOYNA-N
SMILESOS(O)(=O)=O.OS(O)(=O)=O.NCC1OC(OC2C(N)CC(N)C(OC3OC(CO)C(O)C(N)C3O)C2O)C(O)C(O)C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Assay (special)Kanamycin B: ≤4.0%
pH5.5-7.5
Optical Rotation+103.0 to +115.0°
Potency≥670 IU/mg (dried basis)
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only